Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213614.2 | NP_998779.2 | 5 | 371 | ||
GenBank mRNA | AF033350.1 | - | 3 | 218 | ||
AK134700.1 | - | 5 | 308 | |||
BC059848.1 | - | 3 | 249 | |||
BC094347.1 | - | 5 | 344 | |||
BC141073.1 | - | 5 | 381 | 9055685 | ||
BC145331.1 | - | 5 | 381 | 9053887 | ||
BC145332.1 | - | 5 | 394 | 9053888 | ||
CA945297.1 | - | 5 | 387 |