Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213614.2 | NP_998779.2 | 8 | 744 | ||
GenBank mRNA | AF033350.1 | - | 6 | 591 | ||
AK134700.1 | - | 8 | 681 | |||
BC059848.1 | - | 6 | 622 | |||
BC079631.1 | - | 2 | 58 | |||
BC094347.1 | - | 8 | 717 | |||
BC141073.1 | - | 8 | 754 | 9055685 | ||
BC145331.1 | - | 8 | 754 | 9053887 | ||
BC145332.1 | - | 8 | 767 | 9053888 |