Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017462.2 | NP_059490.2 | 14 | 2711 | ||
GenBank mRNA | AB121698.1 | - | 13 | 2404 | ||
AK133584.1 | - | 14 | 2637 | |||
AK148091.1 | - | 14 | 2673 | |||
BC053024.1 | - | 14 | 2637 | |||
BC061472.1 | - | 14 | 2625 | 6827238 | ||
BC080291.1 | - | 14 | 2542 | |||
U53584.1 | - | 14 | 2631 |