Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159496.1 | NP_001152968.1 | 3, 4 | 1365 | ||
NM_001159497.1 | NP_001152969.1 | 3, 4 | 1118 | |||
NM_001159498.1 | NP_001152970.1 | 3, 4 | 1041 | |||
NM_011151.2 | NP_035281.1 | 3, 4 | 1365 | |||
GenBank mRNA | AJ271833.1 | - | 3, 4 | 1343 | ||
AJ271836.1 | - | 3, 4 | 1351 | |||
AK010224.1 | - | 2, 3 | 268 | |||
AK076923.1 | - | 3, 4 | 1125 | |||
AK077136.1 | - | 4, 5 | 1113 | |||
AK136077.1 | - | 3, 4 | 1367 | |||
AK160653.1 | - | 3, 4 | 1366 | |||
AK163283.1 | - | 3, 4 | 1358 | |||
AK167176.1 | - | 3, 4 | 1364 | |||
BC018556.1 | - | 3, 4 | 1056 | 3987825 | ||
BC090963.1 | - | 2, 3 | 956 | 30632168 | ||
D17411.1 | - | 3, 4 | 990 | |||
D17412.1 | - | 3, 4 | 990 | |||
D45859.1 | - | 2, 3 | 237 | |||
D45860.1 | - | 3, 4 | 990 | |||
D45861.1 | - | 2, 3 | 237 | |||
U09218.1 | - | 3, 4 | 1049 |