Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013636.3 | NP_038664.2 | 3 | 539 | ||
GenBank mRNA | AK076009.1 | - | 3 | 476 | ||
AK088229.1 | - | 3 | 483 | |||
AK147131.1 | - | 3 | 541 | |||
AK152422.1 | - | 3 | 502 | |||
AK152625.1 | - | 3 | 478 | |||
AK153363.1 | - | 3 | 352 | |||
AK169336.1 | - | 3 | 499 | |||
AK205397.1 | - | 3 | 253 | |||
AK207485.1 | - | 3 | 253 | |||
AK215346.1 | - | 3 | 253 | |||
AK218771.1 | - | 3 | 253 | |||
BC008157.1 | - | 3 | 457 | |||
BC010613.1 | - | 3 | 312 | 3589168 | ||
BC021646.1 | - | 3 | 540 | 3487479 | ||
BC039972.1 | - | 3 | 472 | |||
BC085496.1 | - | 3 | 478 | 6829867 | ||
D85137.1 | - | 3 | 364 | |||
M27071.1 | - | 3 | 460 | |||
U53456.1 | - | 3 | 456 |
Gene Symbol: Gm5601 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK215748.1 | - | 1 | 254 |