Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011161.5 | NP_035291.4 | 11 | 1064 | ||
GenBank mRNA | AF135185.1 | - | 11 | 1044 | ||
AK144846.1 | - | 11 | 1037 | |||
AK161190.1 | - | 9 | 1239 | |||
AK161562.1 | - | 11 | 1016 | |||
BC057211.1 | - | 11 | 1000 | |||
BC064737.1 | - | 11 | 1015 | 30112690 | ||
BC092526.1 | - | 11 | 1064 | 6593321 |