Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000596.2 | NP_000587.1 | 4 | 954 | ||
GenBank mRNA | AK314925.1 | - | 4 | 826 | ||
AV695706.2 | - | 4 | 277 | |||
BC035263.1 | - | 4 | 815 | 4800940 | ||
BC057806.1 | - | 4 | 826 | 30337849 | ||
BT019685.1 | - | 4 | 661 | |||
FJ795026.1 | - | 4 | 660 | |||
M20841.1 | - | 4 | 781 | |||
M31145.1 | - | 4 | 825 | |||
X12385.1 | - | 4 | 811 | |||
X13405.1 | - | 4 | 763 | |||
X15002.1 | - | 3 | 241 | |||
Y00856.1 | - | 4 | 713 |