Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011188.3 | NP_035318.1 | 6, 7 | 824 | ||
GenBank mRNA | AK005083.1 | - | 5, 6 | 447 | ||
AK076874.1 | - | 6, 7 | 826 | |||
AK146919.1 | - | 5, 6 | 465 | |||
AK153413.1 | - | 5, 6 | 468 | |||
AK168548.1 | - | 5, 6 | 465 | |||
BC005462.1 | - | 5, 6 | 474 | 3590739 | ||
U61283.1 | - | 5, 6 | 510 |