Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011188.3 | NP_035318.1 | 2 | 429 | ||
GenBank mRNA | AK005083.1 | - | 1 | 52 | ||
AK076874.1 | - | 2 | 431 | |||
AK146919.1 | - | 1 | 70 | |||
AK153413.1 | - | 1 | 73 | |||
AK168548.1 | - | 1 | 70 | |||
BC005462.1 | - | 1 | 79 | 3590739 |