Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008950.1 | NP_032976.1 | 5 | 447 | ||
GenBank mRNA | AK010505.1 | - | 5 | 467 | ||
AK012294.1 | - | 5 | 480 | |||
AK135451.1 | - | 5 | 457 | |||
AK145703.1 | - | 5 | 451 | |||
BC004052.1 | - | 1 | 142 | |||
BC030840.1 | - | 5 | 479 | 3994523 | ||
Z54219.1 | - | 5 | 447 |