Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008973.2 | NP_032999.1 | 4, 5 | 1923 | ||
GenBank mRNA | AK011346.1 | - | 4, 5 | 780 | ||
AK011699.1 | - | 4, 5 | 706 | |||
AK161964.1 | - | 4, 5 | 614 | |||
BC002064.1 | - | 4, 5 | 492 | 3488877 | ||
BC061695.1 | - | 4, 5 | 723 | 6313227 | ||
D90225.1 | - | 4, 5 | 667 | |||
EF437379.1 | - | 1, 2 | 154 |