Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGSF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170961.1 | NP_001164432.1 | 3, 4 | 373 | ||
NM_001170963.1 | NP_001164434.1 | 3, 4 | 197 | |||
NM_001555.4 | NP_001546.2 | 3, 4 | 373 | |||
NM_205833.3 | NP_991402.1 | 3, 4 | 373 | |||
GenBank mRNA | AB058894.2 | - | 3, 4 | 225 | ||
AK057916.1 | - | 3, 4 | 204 | |||
AK226008.1 | - | 3, 4 | 230 | |||
AK307871.1 | - | 3, 4 | 249 | |||
BC063884.1 | - | 3, 4 | 209 | 30387876 | ||
Y10523.1 | - | 2, 3 | 170 |