Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvalb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013645.3 | NP_038673.2 | 3 | 258 | ||
GenBank mRNA | AK002354.1 | - | 3 | 266 | ||
AK005371.1 | - | 3 | 256 | |||
AK013561.1 | - | 3 | 250 | |||
AK131986.1 | - | 3 | 328 | |||
BC027424.1 | - | 3 | 258 | 4925213 | ||
S75909.1 | - | 4 | 319 | |||
S75910.1 | - | 1195 | ||||
X54613.1 | - | 3 | 236 | |||
X59382.1 | - | 4 | 292 |