Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008990.3 | NP_033016.3 | 9 | 1773 | ||
GenBank mRNA | BC046770.1 | - | 9 | 1669 | 5703547 | |
BC059941.1 | - | 9 | 1725 | 6530953 | ||
D26107.1 | - | 9 | 1502 |