Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvrl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159724.1 | NP_001153196.1 | 4, 5 | 1129 | ||
NM_008990.3 | NP_033016.3 | 4, 5 | 1129 | |||
GenBank mRNA | AK075815.1 | - | 4, 5 | 1034 | ||
BC009088.1 | - | 4, 5 | 1022 | 2812478 | ||
BC046770.1 | - | 4, 5 | 1025 | 5703547 | ||
BC059941.1 | - | 4, 5 | 1081 | 6530953 | ||
D26107.1 | - | 4, 5 | 858 | |||
M80206.1 | - | 4, 5 | 858 |