Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159525.1 | NP_001152997.1 | 4 | 469 | ||
NM_023041.3 | NP_075528.3 | 6 | 745 | |||
GenBank mRNA | AK012785.1 | - | 6 | 728 | ||
AK028449.1 | - | 4 | 463 | |||
AK029368.1 | - | 6 | 746 | |||
AK075576.1 | - | 6 | 732 | |||
AK170175.1 | - | 6 | 734 | |||
AK181194.1 | - | 1 | 123 | |||
AK210467.1 | - | 1 | 105 | |||
AK216660.1 | - | 1 | 123 | |||
AK218946.1 | - | 3 | 327 | |||
BC012517.1 | - | 6 | 680 | |||
BC019767.1 | - | 6 | 728 | 4502819 | ||
CT010305.1 | - | 6 | 721 | |||
Y09046.1 | - | 6 | 726 |