Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011226.1 | NP_035356.1 | 2 | 145 | ||
GenBank mRNA | AB232613.1 | - | 1 | 51 | ||
AK017590.1 | - | 2 | 179 | |||
BC032936.1 | - | 2 | 145 | 1379043 | ||
M79309.1 | - | 1 | 51 | |||
X80473.1 | - | 1 | 51 |