Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029391.2 | NP_083667.1 | 6, 7 | 665 | ||
GenBank mRNA | AB232592.1 | - | 6, 7 | 524 | ||
AK005314.1 | - | 6, 7 | 665 | |||
AK140513.1 | - | 5, 6 | 862 | |||
AK170552.1 | - | 6, 7 | 679 | |||
AK180450.1 | - | 1, 2 | 175 | |||
AK219531.1 | - | 2, 3 | 214 | |||
BC007147.1 | - | 6, 7 | 609 | 3600193 | ||
BC096654.1 | - | 6, 7 | 648 | 5056766 |