Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166406.1 | NP_001159878.1 | 12 | 1720 | ||
NM_001166407.1 | NP_001159879.1 | 12 | 1718 | |||
NM_009004.4 | NP_033030.1 | 12 | 1614 | |||
GenBank mRNA | AK051201.1 | - | 12 | 1532 | ||
AK051995.1 | - | 12 | 1569 | |||
AK053280.1 | - | 12 | 1533 | |||
AK083412.1 | - | 12 | 1574 | |||
AK084732.1 | - | 12 | 1556 | |||
AK142764.1 | - | 12 | 1562 | |||
AK144844.1 | - | 12 | 1532 | |||
BC060608.1 | - | 12 | 1720 | 5720408 | ||
Y09632.1 | - | 12 | 1769 |