Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011237.2 | NP_035367.1 | 2 | 144 | ||
GenBank mRNA | AF045663.1 | - | 2 | 144 | ||
AK134293.1 | - | 2 | 157 | |||
AK138973.1 | - | 2 | 149 | |||
AK165833.1 | - | 2 | 179 | |||
BC082556.1 | - | 2 | 83 | 30538342 |