Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146174.1 | NP_001139646.1 | 13 | 1657 | ||
NM_011241.4 | NP_035371.4 | 14 | 1705 | |||
GenBank mRNA | AK042930.1 | - | 13 | 1623 | ||
AK129452.1 | - | 13 | 1586 | |||
AK133905.1 | - | 13 | 1625 | |||
AK145459.1 | - | 12 | 1260 | |||
AK146670.1 | - | 14 | 1665 | |||
AK159009.1 | - | 13 | 1610 | |||
AK159525.1 | - | 13 | 1622 | |||
AK159930.1 | - | 13 | 1613 | |||
AK177694.1 | - | 3 | 188 | |||
AK179704.1 | - | 3 | 189 | |||
AK186071.1 | - | 3 | 189 | |||
BC014855.1 | - | 13 | 1613 | 3157225 | ||
BC052862.1 | - | 13 | 1580 | 30063509 | ||
BC066213.1 | - | 13 | 1621 | 30466595 | ||
BC071200.1 | - | 14 | 1821 | 5032610 | ||
U08110.1 | - | 14 | 1688 |