Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146174.1 | NP_001139646.1 | 2 | 331 | ||
NM_011241.4 | NP_035371.4 | 3 | 379 | |||
GenBank mRNA | AK035571.1 | - | 3 | 341 | ||
AK042930.1 | - | 2 | 296 | |||
AK088768.1 | - | 2 | 253 | |||
AK129452.1 | - | 2 | 260 | |||
AK133905.1 | - | 2 | 296 | |||
AK145459.1 | - | 2 | 252 | |||
AK146670.1 | - | 3 | 339 | |||
AK159009.1 | - | 2 | 284 | |||
AK159525.1 | - | 2 | 296 | |||
AK159930.1 | - | 2 | 287 | |||
BC014855.1 | - | 2 | 287 | 3157225 | ||
BC052862.1 | - | 2 | 254 | 30063509 | ||
BC066213.1 | - | 2 | 295 | 30466595 | ||
BC071200.1 | - | 3 | 495 | 5032610 | ||
U08110.1 | - | 3 | 362 |