Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009025.2 | NP_033051.2 | 16 | 1680 | ||
GenBank mRNA | AK053353.1 | - | 16 | 1676 | ||
AK075768.1 | - | 2 | 318 | |||
AK141511.1 | - | 16 | 1687 | |||
AK147896.1 | - | 16 | 1648 | |||
AK166354.1 | - | 16 | 1678 | |||
BC057300.1 | - | 16 | 1671 | 6821254 | ||
BC060195.1 | - | 16 | 1695 | |||
BC068297.1 | - | 16 | 1544 | 30102545 | ||
U20238.1 | - | 16 | 1665 |