Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166623.1 | NP_001160095.1 | 6 | 856 | ||
NM_011252.4 | NP_035382.1 | 6 | 853 | |||
GenBank mRNA | AJ237846.1 | - | 6 | 783 | ||
AJ237847.1 | - | 6 | 798 | |||
AK032846.1 | - | 1 | 74 | |||
AK083899.1 | - | 6 | 854 | |||
AK135164.1 | - | 6 | 858 | |||
AK165006.1 | - | 6 | 799 | |||
AK178027.1 | - | 2 | 195 | |||
AK211002.1 | - | 1 | 68 | |||
BC003710.1 | - | 6 | 851 | 3599678 |