Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF169300.1 | - | 8 | 783 | ||
AK019171.1 | - | 8 | 798 | |||
AK019656.1 | - | 8 | 834 | |||
AK037465.1 | - | 198 | ||||
AK090162.1 | - | 8 | 821 | |||
AK145596.1 | - | 8 | 760 | |||
AK147057.1 | - | 8 | 743 | |||
AK151116.1 | - | 8 | 791 | |||
AK158987.1 | - | 8 | 800 | |||
AK159296.1 | - | 8 | 798 | |||
AK159778.1 | - | 8 | 792 | |||
AK159949.1 | - | 8 | 840 | |||
AK217419.1 | - | 1 | 39 | |||
BC003282.1 | - | 8 | 762 | 3582133 | ||
U95141.1 | - | 6 | 473 |