Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011293.2 | NP_035423.2 | 2 | 138 | ||
GenBank mRNA | AK158610.1 | - | 2 | 139 | ||
AK158644.1 | - | 2 | 128 | |||
BC042939.1 | - | 2 | 95 | 6476299 | ||
D85818.1 | - | 2 | 92 |