Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009095.2 | NP_033121.2 | 5 | 568 | ||
GenBank mRNA | AK002235.1 | - | 5 | 562 | ||
AK009663.1 | - | 5 | 562 | |||
AK010681.1 | - | 5 | 562 | |||
AK012424.1 | - | 5 | 562 | |||
AK012481.1 | - | 5 | 562 | |||
AK020477.1 | - | 5 | 561 | |||
AK020754.1 | - | 5 | 562 | |||
AK050611.1 | - | 5 | 562 | |||
AK050622.1 | - | 5 | 562 | |||
AK152449.1 | - | 5 | 562 | |||
AK160552.1 | - | 5 | 562 | |||
AK164079.1 | - | 5 | 562 | |||
AK167533.1 | - | 5 | 562 | |||
AK168044.1 | - | 5 | 562 | |||
AK168106.1 | - | 5 | 562 | |||
AK169016.1 | - | 5 | 562 | |||
AK172446.1 | - | 5 | 562 | |||
BC020401.1 | - | 5 | 539 | |||
BC058690.1 | - | 5 | 549 | 6814814 | ||
U78085.1 | - | 5 | 554 | |||
Y12431.1 | - | 5 | 544 |