Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009096.3 | NP_033122.1 | 5 | 1260 | ||
GenBank mRNA | AK146952.1 | - | 5 | 1261 |