Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011299.4 | NP_035429.1 | 17 | 1841 | ||
GenBank mRNA | AJ131021.1 | - | 17 | 1669 | ||
AK019881.1 | - | 17 | 1746 | |||
BC038251.1 | - | 11 | 1068 | |||
BC043064.1 | - | 17 | 1791 | 5706153 | ||
BC051079.1 | - | 17 | 1751 | 6491001 | ||
BC055331.1 | - | 17 | 1841 | 6406879 | ||
BC056946.1 | - | 17 | 1784 | 6822831 | ||
X57237.1 | - | 1 | 13 |