Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172424.1 | NP_001165895.1 | 6, 7 | 830 | ||
NM_011303.6 | NP_035433.1 | 5, 6 | 1194 | |||
GenBank mRNA | AF061743.1 | - | 5, 6 | 996 | ||
AK032958.1 | - | 1257 | ||||
AK087137.1 | - | 5, 6 | 1270 | |||
AK151419.1 | - | 5, 6 | 1195 | |||
BC008980.1 | - | 5, 6 | 710 | 3587135 | ||
BC010972.1 | - | 5, 6 | 1223 | 4215444 | ||
BC013540.1 | - | 5, 6 | 1173 | 4241608 | ||
X95281.1 | - | 5, 6 | 1232 |