Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rtn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025364.3 | NP_001020535.1 | 2 | 267 | ||
NM_013648.6 | NP_038676.1 | 5 | 1181 | |||
GenBank mRNA | AF038538.1 | - | 5 | 1137 | ||
AF038539.1 | - | 2 | 206 | |||
AK144271.1 | - | 5 | 1144 | |||
AK160500.1 | - | 2 | 228 | |||
BC031370.1 | - | 5 | 1138 | 5359358 | ||
W65979.1 | - | 2 | 267 |