Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sar1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009120.2 | NP_033146.1 | 6 | 530 | ||
GenBank mRNA | AK157843.1 | - | 6 | 542 | ||
AK159239.1 | - | 6 | 541 | |||
AK168175.1 | - | 6 | 542 | |||
AK192256.1 | - | 1 | 44 | |||
BC005549.1 | - | 6 | 531 | 3585667 | ||
L20294.1 | - | 6 | 505 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK129305.1 | - | 3728 | Chr. - - [-] on Build |