Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204979.1 | NP_001191908.1 | 6, 7 | 868 | ||
NM_011319.3 | NP_035449.1 | 6, 7 | 868 | |||
GenBank mRNA | AK028358.1 | - | 6, 7 | 800 | ||
AK075827.1 | - | 6, 7 | 799 | |||
AK082793.1 | - | 6, 7 | 779 | |||
AK152150.1 | - | 6, 7 | 826 | |||
AK153158.1 | - | 6, 7 | 818 | |||
AK181805.1 | - | 2, 3 | 274 | |||
AK184943.1 | - | 2, 3 | 273 | |||
AK190309.1 | - | 2, 3 | 273 | |||
AK217331.1 | - | 2, 3 | 273 | |||
AK219731.1 | - | 2, 3 | 274 | |||
BC008612.1 | - | 6, 7 | 757 | 3257101 | ||
BC031932.1 | - | 6, 7 | 757 | |||
M74012.1 | - | 6, 7 | 820 |