Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scn8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077499.2 | NP_001070967.1 | 15 | 2605 | ||
NM_011323.3 | NP_035453.2 | 14 | 2506 | |||
GenBank mRNA | AF049617.1 | - | 14 | 2452 | ||
AK083220.1 | - | 1 | 103 | |||
U26707.1 | - | 12 | 1726 |