Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sct | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287171.1 | NP_001274100.1 | 2 | 137 | ||
NM_011328.3 | NP_035458.1 | 2 | 137 | |||
GenBank mRNA | AK145859.1 | - | 2 | 136 | ||
AK167408.1 | - | 2 | 136 | |||
BC048484.1 | - | 2 | 137 | 6707701 | ||
CF616255.1 | - | 115 | ||||
X73580.1 | - | 2 | 110 |