Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021443.3 | NP_067418.1 | 2 | 203 | ||
GenBank mRNA | AB023418.1 | - | 2 | 211 | ||
AK007942.1 | - | 2 | 204 | |||
BC117101.1 | - | 2 | 201 | 40126420 | ||
BC117103.1 | - | 2 | 201 | 40126422 | ||
BY707942.1 | - | 2 | 204 |