Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009141.3 | NP_033167.2 | 2 | 344 | ||
GenBank mRNA | AK044397.1 | - | 1 | 411 | ||
AK156787.1 | - | 1 | 414 | |||
BC024392.1 | - | 2 | 256 | 4976854 | ||
BQ829929.1 | - | 2 | 262 | |||
U27267.1 | - | 2 | 264 |