Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000641.3 | NP_000632.1 | 5 | 1151 | ||
NM_001267718.1 | NP_001254647.1 | 4 | 978 | |||
GenBank mRNA | BC012506.1 | - | 5 | 1134 | 4476678 | |
M57765.1 | - | 5 | 1134 | |||
X58377.1 | - | 5 | 1061 |