Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009178.3 | NP_033204.2 | 7 | 585 | ||
GenBank mRNA | AB061305.1 | - | 7 | 533 | ||
AK008543.1 | - | 1 | 63 | |||
AK131874.1 | - | 1 | 63 | |||
BC011121.1 | - | 7 | 530 | 3497996 | ||
BC050773.1 | - | 7 | 585 | 6587379 | ||
D28941.1 | - | 7 | 540 | |||
X95809.1 | - | 6 | 412 |