Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276425.1 | NP_001263354.1 | 5 | 942 | ||
NM_011373.3 | NP_035503.1 | 4 | 1295 | |||
GenBank mRNA | AJ007310.1 | - | 5 | 937 | ||
AK007601.1 | - | 5 | 924 | |||
AK085730.1 | - | 5 | 933 | |||
AK144884.1 | - | 5 | 933 | |||
AK153036.1 | - | 5 | 902 | |||
AK155956.1 | - | 4 | 2224 | |||
AK159712.1 | - | 5 | 1286 | |||
AK161559.1 | - | 5 | 927 | |||
BC056451.1 | - | 4 | 748 | 5685866 | ||
BC086784.1 | - | 883 | 6844232 | |||
Y15779.1 | - | 5 | 919 | |||
Y15780.1 | - | 4 | 931 |