Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009185.3 | NP_033211.2 | 12 | 1568 | ||
GenBank mRNA | AK049223.1 | - | 12 | 1505 | ||
BC049865.1 | - | 12 | 1522 | |||
BC060706.1 | - | 13 | 1590 | |||
BC087545.1 | - | 11 | 1259 | |||
BC108373.1 | - | 11 | 1236 | |||
BC141175.1 | - | 12 | 1441 | 9055787 | ||
BC145493.1 | - | 12 | 1441 | 9054056 | ||
U36778.1 | - | 12 | 1486 |