Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL17A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002190.2 | NP_002181.1 | 2 | 218 | ||
GenBank mRNA | BC066251.1 | - | 2 | 216 | 6971875 | |
BC066252.1 | - | 2 | 216 | 6971876 | ||
BC066253.1 | - | 2 | 216 | 6971878 | ||
BC067503.1 | - | 2 | 216 | 6971877 | ||
BC067504.1 | - | 2 | 216 | 6971879 | ||
BC067505.1 | - | 2 | 216 | 6971881 | ||
HM535222.1 | - | 1 | 37 | |||
U32659.1 | - | 2 | 226 | |||
Z58820.1 | - | 2 | 215 |