Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175090.4 | NP_780299.2 | 4 | 489 | ||
GenBank mRNA | AK046835.1 | - | 4 | 442 | ||
AK151337.1 | - | 4 | 471 | |||
AK170285.1 | - | 4 | 442 | |||
BC034674.1 | - | 4 | 427 | 3968084 | ||
BC046470.1 | - | 2 | 163 | |||
BC058227.1 | - | 4 | 429 | 6309323 | ||
BC065147.1 | - | 3 | 337 |