Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc31a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175090.4 | NP_780299.2 | 2, 3 | 338 | ||
GenBank mRNA | AK046835.1 | - | 2, 3 | 291 | ||
AK151337.1 | - | 2, 3 | 320 | |||
AK170285.1 | - | 2, 3 | 291 | |||
AK203972.1 | - | 2, 3 | 214 | |||
BC034674.1 | - | 2, 3 | 276 | 3968084 | ||
BC046470.1 | - | 12 | ||||
BC058227.1 | - | 2, 3 | 278 | 6309323 | ||
BC065147.1 | - | 1, 2 | 186 | |||
BY097629.1 | - | 2, 3 | 339 |