Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarca4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174078.1 | NP_001167549.1 | 29 | 4542 | ||
NM_001174079.1 | NP_001167550.1 | 29 | 4533 | |||
NM_011417.3 | NP_035547.2 | 29 | 4533 | |||
GenBank mRNA | AK147047.1 | - | 14 | 1849 | ||
AK147285.1 | - | 29 | 4487 | |||
AK160825.1 | - | 29 | 4446 | |||
AK166837.1 | - | 29 | 4438 | |||
BC023186.1 | - | 8 | 1122 | |||
BC026672.1 | - | 12 | 1682 | |||
BC060229.1 | - | 29 | 4427 | |||
BC079560.1 | - | 29 | 4533 | 30533489 | ||
S68108.1 | - | 19 | 2499 |