Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncor2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253904.1 | NP_001240833.1 | 21 | 2435 | ||
NM_001253905.1 | NP_001240834.1 | 17 | 1989 | |||
NM_011424.3 | NP_035554.3 | 21 | 2600 | |||
GenBank mRNA | AF113002.1 | - | 17 | 1981 | ||
AF125671.1 | - | 21 | 2435 | |||
AK147394.1 | - | 21 | 2401 |