siRNA Details
Gene Symbol: Snrpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082961.1 | NP_001076430.1 | 6 | 676 | ||
NM_001082962.1 | NP_001076431.1 | 6 | 720 | |||
NM_013670.3 | NP_038698.1 | 5 | 614 | |||
GenBank mRNA | AK010671.1 | - | 5 | 651 | ||
AK013607.1 | - | 5 | 571 | |||
AK144386.1 | - | 5 | 570 | |||
BC019589.1 | - | 5 | 562 | 3484323 | ||
BC024880.1 | - | 5 | 553 | 3987218 | ||
BC069894.1 | - | 5 | 551 | |||
BQ042870.1 | - | 6 | 678 | |||
S40304.1 | - | 2 | 259 | |||
S62288.1 | - | 2 | 136 | |||
X62648.1 | - | 2 | 259 | |||
X63730.1 | - | 2 | 212 |
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033174.3 | NP_149409.1 | 5 | 614 | ||
GenBank mRNA | CF751578.1 | - | 1 | 113 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -




