Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009228.2 | NP_033254.2 | 2 | 427 | ||
GenBank mRNA | AK037183.1 | - | 2 | 325 | ||
AK052932.1 | - | 1 | 2721 | |||
AK137379.1 | - | 2 | 428 | |||
AK181361.1 | - | 1 | 17 | |||
BB625367.1 | - | 2 | 420 | |||
BC018546.1 | - | 2 | 377 | 3674921 | ||
U00677.1 | - | 2 | 424 |