siRNA Details
Gene Symbol: Sox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011443.3 | NP_035573.3 | 1 | 2148 | ||
GenBank mRNA | AK131933.1 | - | 1 | 1583 | ||
BC057574.1 | - | 2 | 1986 | 6413283 | ||
U31967.1 | - | 3 | 1976 |

More Information
Other Species Targeted

Panther Classification:
