Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018042.3 | NP_001018052.1 | 4 | 636 | ||
NM_001098425.1 | NP_001091895.1 | 2 | 545 | |||
GenBank mRNA | AF062567.1 | - | 2 | 272 | ||
AK004607.1 | - | 1 | 286 | |||
AK088238.1 | - | 2 | 520 | |||
AK132702.1 | - | 2 | 291 | |||
BC079874.1 | - | 4 | 636 | 6833706 | ||
CA328978.1 | - | 2 | 549 |